ADVH49B820C3 AChemBlock

5,6,7,8-tetrahydro-1,7-naphthyridin-8-one

  • Cas Number : 301666-63-7
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-6-7-8-tetrahydro-1-7-naphthyridin-8-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76226
Availability details for
Quantity
  • Availability
  • Add to Cart