ADVH49B820C4 AChemBlock

2,3-dihydro-1H-indene-5-carbaldehyde

  • Cas Number : 30084-91-4
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-dihydro-1h-indene-5-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76230
Availability details for
Quantity
  • Availability
  • Add to Cart