ADVH49B820C6 AChemBlock

(1S)-1-(3-nitrophenyl)ethanamine

  • Cas Number : 297730-25-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-1-3-nitrophenyl-ethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76246
Availability details for
Quantity
  • Availability
  • Add to Cart