ADVH49B820CB AChemBlock

2,6-dichloro-3-nitropyridin-4-amine

  • Cas Number : 2897-43-0
  • Molecular Weight : 208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloro-3-nitropyridin-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71468
Availability details for
Quantity
  • Availability
  • Add to Cart