ADVH49B820D1 AChemBlock

5-bromo-2,4-dimethylthiazole

  • Cas Number : 28599-52-2
  • Molecular Weight : 192.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-4-dimethylthiazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71470
Availability details for
Quantity
  • Availability
  • Add to Cart