ADVH49B820D2 AChemBlock

2-methyl-9H-fluoren-9-one

  • Cas Number : 2840-51-9
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-9h-fluoren-9-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71473
Availability details for
Quantity
  • Availability
  • Add to Cart