ADVH49B820D4 AChemBlock

4-fluoro-1,2,3,6-tetrahydropyridine;hydrochloride

  • Cas Number : 281652-11-7
  • Molecular Weight : 137.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1-2-3-6-tetrahydropyridine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76302
Availability details for
Quantity
  • Availability
  • Add to Cart