ADVH49B820D6 AChemBlock

2-tert-butyl-2H-1,2,3,4-tetrazole-5-carboxylic acid

  • Cas Number : 1547646-28-5
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butyl-2h-1-2-3-4-tetrazole-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16136
Availability details for
Quantity
  • Availability
  • Add to Cart