ADVH49B820E1 AChemBlock

tert-butyl 4-(ethylamino)piperidine-1-carboxylate

  • Cas Number : 264905-39-7
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-ethylamino-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71486
Availability details for
Quantity
  • Availability
  • Add to Cart