ADVH49B820E8 AChemBlock

5-methyl-1,3-thiazole-4-carbaldehyde

  • Cas Number : 261710-79-6
  • Molecular Weight : 127.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1-3-thiazole-4-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76391
Availability details for
Quantity
  • Availability
  • Add to Cart