ADVH49B820EA AChemBlock

methyl 2-(4-oxo-3,4-dihydrophthalazin-1-yl)acetate

  • Cas Number : 25947-14-2
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-4-oxo-3-4-dihydrophthalazin-1-yl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76410
Availability details for
Quantity
  • Availability
  • Add to Cart