ADVH49B820ED AChemBlock

6-Chloro-1H-indole-5-carboxylic acid

  • Cas Number : 256935-86-1
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1h-indole-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72396
Availability details for
Quantity
  • Availability
  • Add to Cart