ADVH49B820EE AChemBlock

tert-butyl 4-[2-(hydroxymethyl)phenyl]piperidine-1-carboxylate

  • Cas Number : 255051-62-8
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-2-hydroxymethyl-phenyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76431
Availability details for
Quantity
  • Availability
  • Add to Cart