ADVH49B820F0 AChemBlock

N-Boc-3,5-dichloroaniline

  • Cas Number : 25216-60-8
  • Molecular Weight : 262.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-3-5-dichloroaniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70348
Availability details for
Quantity
  • Availability
  • Add to Cart