ADVH49B820F8 AChemBlock

2,3-dihydro-6-nitroinden-1-one

  • Cas Number : 24623-24-3
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-dihydro-6-nitroinden-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71498
Availability details for
Quantity
  • Availability
  • Add to Cart