ADVH49B820F9 AChemBlock

Methyl (4-bromo-2-formylphenoxy)acetate

  • Cas Number : 24581-99-5
  • Molecular Weight : 273.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-2-formylphenoxy-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86726
Availability details for
Quantity
  • Availability
  • Add to Cart