ADVH49B820FD AChemBlock

7-hydroxy-1H-indole-3-carboxylic acid

  • Cas Number : 24370-79-4
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-hydroxy-1h-indole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76485
Availability details for
Quantity
  • Availability
  • Add to Cart