ADVH49B8210B AChemBlock

1H,2H,3H-pyrrolo[3,2-c]pyridine

  • Cas Number : 23596-28-3
  • Molecular Weight : 120.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1h-2h-3h-pyrrolo-3-2-c-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76528
Availability details for
Quantity
  • Availability
  • Add to Cart