ADVH49B8210C AChemBlock

3-chloro-6-methoxy-2-methyl-benzoic acid

  • Cas Number : 23550-92-7
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-6-methoxy-2-methyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76533
Availability details for
Quantity
  • Availability
  • Add to Cart