ADVH49B82110 AChemBlock

TR-14035

  • Cas Number : 232271-19-1
  • Molecular Weight : 474.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/tr-14035.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86768
Availability details for
Quantity
  • Availability
  • Add to Cart