ADVH49B82112 AChemBlock

2-methylindazole-4-carboxamide

  • Cas Number : 2306264-94-6
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methylindazole-4-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76556
Availability details for
Quantity
  • Availability
  • Add to Cart