ADVH49B82114 AChemBlock

(1S)-2,2-difluorocyclopropanamine;hydrochloride

  • Cas Number : 2306252-66-2
  • Molecular Weight : 129.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2-2-difluorocyclopropanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76558
Availability details for
Quantity
  • Availability
  • Add to Cart