ADVH49B82117 AChemBlock

5-chloro-2-methoxybenzene-1-sulfonyl chloride

  • Cas Number : 22952-32-5
  • Molecular Weight : 241.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-2-methoxybenzene-1-sulfonyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71514
Availability details for
Quantity
  • Availability
  • Add to Cart