ADVH49B82119 AChemBlock

Acetone-[bis-(2-amino-ethyl)-dithioacetal]

  • Cas Number : 22907-30-8
  • Molecular Weight : 194.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/acetone-bis-2-amino-ethyl-dithioacetal.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86701
Availability details for
Quantity
  • Availability
  • Add to Cart