ADVH49B8212C AChemBlock

2-(2-trimethylsilylethynyl)pyrimidin-5-amine

  • Cas Number : 2243974-50-5
  • Molecular Weight : 191.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-trimethylsilylethynyl-pyrimidin-5-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76594
Availability details for
Quantity
  • Availability
  • Add to Cart