ADVH49B82134 AChemBlock

2-{bis[(tert-butoxy)carbonyl]amino}pyrimidine-5-carboxylic acid

  • Cas Number : 2231168-34-4
  • Molecular Weight : 339.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-bis-tert-butoxy-carbonyl-amino-pyrimidine-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76610
Availability details for
Quantity
  • Availability
  • Add to Cart