ADVH49B8213A AChemBlock

methyl 1-[(3R)-3-{[(tert-butoxy)carbonyl]amino}-4-(phenylsulfanyl)butyl]piperidine-4-carboxylate; oxalic acid

  • Cas Number : 2227199-29-1
  • Molecular Weight : 512.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-3r-3-tert-butoxy-carbonyl-amino-4-phenylsulfanyl-butyl-piperidine-4-carboxylate-oxalic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76650
Availability details for
Quantity
  • Availability
  • Add to Cart