ADVH49B82140 AChemBlock

2-((6-chloropyrimidin-4-yl)amino)ethanol

  • Cas Number : 22177-94-2
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-6-chloropyrimidin-4-yl-amino-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86514
Availability details for
Quantity
  • Availability
  • Add to Cart