ADVH49B82141 AChemBlock

4-chloro-6-fluoro-7-methyl-quinoline

  • Cas Number : 2216746-75-5
  • Molecular Weight : 195.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-6-fluoro-7-methyl-quinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76690
Availability details for
Quantity
  • Availability
  • Add to Cart