ADVH49B82142 AChemBlock

4-Isopropoxy-4'-nitrobenzophenone

  • Cas Number : 221532-00-9
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-isopropoxy-4-nitrobenzophenone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86704
Availability details for
Quantity
  • Availability
  • Add to Cart