ADVH49B82144 AChemBlock

4-(trifluoromethyl)pyrimidine-5-carboxylic acid

  • Cas Number : 220880-12-6
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-trifluoromethyl-pyrimidine-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76710
Availability details for
Quantity
  • Availability
  • Add to Cart