ADVH49B82145 AChemBlock

tert-butyl 4-chloropyridine-2-carboxylate

  • Cas Number : 220000-86-2
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-chloropyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76733
Availability details for
Quantity
  • Availability
  • Add to Cart