ADVH49B82147 AChemBlock

tert-butyl (3R)-3-amino-3-methyl-piperidine-1-carboxylate

  • Cas Number : 2199214-46-3
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-3-amino-3-methyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76738
Availability details for
Quantity
  • Availability
  • Add to Cart