ADVH49B82152 AChemBlock

[3-[(3S,4S)-4-amino-3-methyl-2-oxa-8-azaspiro[4.5]decan-8-yl]-6-(2,3-dichlorophenyl)-5-methyl-pyrazin-2-yl]methanol

  • Cas Number : 2172651-73-7
  • Molecular Weight : 437.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-3s-4s-4-amino-3-methyl-2-oxa-8-azaspiro-4-5-decan-8-yl-6-2-3-dichlorophenyl-5-methyl-pyrazin-2-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76809
Availability details for
Quantity
  • Availability
  • Add to Cart