ADVH49B82154 AChemBlock

1-methyl-2-oxabicyclo[2.1.1]hexan-4-amine;hydrochloride

  • Cas Number : 2170372-24-2
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-2-oxabicyclo-2-1-1-hexan-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76834
Availability details for
Quantity
  • Availability
  • Add to Cart