ADVH49B82157 AChemBlock

1-ethyl-4-oxo-quinoline-3-sulfonyl chloride

  • Cas Number : 2169907-32-6
  • Molecular Weight : 271.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-ethyl-4-oxo-quinoline-3-sulfonyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76841
Availability details for
Quantity
  • Availability
  • Add to Cart