ADVH49B8215C AChemBlock

2-[(2S)-piperazin-2-yl]acetonitrile dihydrochloride

  • Cas Number : 2158301-19-8
  • Molecular Weight : 198.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2s-piperazin-2-yl-acetonitrile-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76928
Availability details for
Quantity
  • Availability
  • Add to Cart