ADVH49B8215F AChemBlock

[(1S,2R)-2-aminocyclohexyl]methanol

  • Cas Number : 213993-30-7
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2r-2-aminocyclohexyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76962
Availability details for
Quantity
  • Availability
  • Add to Cart