ADVH49B82164 AChemBlock

1,2-di([1,1'-biphenyl]-4-yl)ethyne

  • Cas Number : 21326-80-7
  • Molecular Weight : 330.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-di-1-1-biphenyl-4-yl-ethyne.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86518
Availability details for
Quantity
  • Availability
  • Add to Cart