ADVH49B82165 AChemBlock

4,6-dichloro-5-fluoropyrimidine

  • Cas Number : 213265-83-9
  • Molecular Weight : 166.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-5-fluoropyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71531
Availability details for
Quantity
  • Availability
  • Add to Cart