ADVH49B8216A AChemBlock

Trans-[4-(methylamino)cyclohexyl]methanesulfonic acid

  • Cas Number : 2124221-12-9
  • Molecular Weight : 207.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-4-methylamino-cyclohexyl-methanesulfonic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76999
Availability details for
Quantity
  • Availability
  • Add to Cart