ADVH49B8216C AChemBlock

2-(3-pyridyl)propan-2-amine

  • Cas Number : 99980-40-2
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-pyridyl-propan-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72454
Availability details for
Quantity
  • Availability
  • Add to Cart