ADVH49B8216E AChemBlock

methyl 1-[6-(trifluoromethyl)-3-pyridyl]cyclopropanecarboxylate

  • Cas Number : 2119721-57-0
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-6-trifluoromethyl-3-pyridyl-cyclopropanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77005
Availability details for
Quantity
  • Availability
  • Add to Cart