ADVH49B82170 AChemBlock

4-fluoro-3-nitrophenol

  • Cas Number : 2105-96-6
  • Molecular Weight : 157.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-3-nitrophenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71537
Availability details for
Quantity
  • Availability
  • Add to Cart