ADVH49B82172 AChemBlock

methyl 2-(benzyloxycarbonylamino)cyclopropanecarboxylate

  • Cas Number : 2104635-40-5
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-benzyloxycarbonylamino-cyclopropanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77029
Availability details for
Quantity
  • Availability
  • Add to Cart