ADVH49B82173 AChemBlock

ethyl 2-methyl-6,7-dihydro-5H-pyrrolizine-3-carboxylate

  • Cas Number : 2104220-54-2
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-methyl-6-7-dihydro-5h-pyrrolizine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77031
Availability details for
Quantity
  • Availability
  • Add to Cart