ADVH49B82176 AChemBlock

3-amino-N-methyl-oxetane-3-carboxamide

  • Cas Number : 2098000-37-2
  • Molecular Weight : 130.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-amino-n-methyl-oxetane-3-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77048
Availability details for
Quantity
  • Availability
  • Add to Cart