ADVH49B82177 AChemBlock

1-(4-chloro-3-fluoro-2-iodo-phenyl)-4-(trifluoromethyl)triazole

  • Cas Number : 2097065-63-7
  • Molecular Weight : 391.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-chloro-3-fluoro-2-iodo-phenyl-4-trifluoromethyl-triazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77054
Availability details for
Quantity
  • Availability
  • Add to Cart