ADVH49B82179 AChemBlock

2-Fluoro-3-(morpholinomethyl)phenylboronic acid pinacol ester

  • Cas Number : 2096340-24-6
  • Molecular Weight : 339.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-3-morpholinomethyl-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70830
Availability details for
Quantity
  • Availability
  • Add to Cart