ADVH49B8217B AChemBlock

2'-oxo-1',2'-dihydrospiro[cyclopropane-1,3'-indole]-5'-carbonitrile

  • Cas Number : 2093433-63-5
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-oxo-1-2-dihydrospiro-cyclopropane-1-3-indole-5-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77069
Availability details for
Quantity
  • Availability
  • Add to Cart